Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163579.1 | NP_001157051.1 | 17 - 18 | 2970 | 112 | |
NM_001163580.1 | NP_001157052.1 | 17 - 18 | 2922 | 112 | ||
NM_028915.3 | NP_083191.2 | 17 - 18 | 2970 | 112 | ||
GenBank mRNA | AK004646.1 | - | 17 - 18 | 2971 | 112 | |
AK029563.1 | - | 17 - 18 | 2888 | 112 | ||
AK030800.1 | - | 9 - 10 | 1564 | 112 | ||
AK078433.1 | - | 6 - 7 | 967 | 112 | ||
AK173256.1 | - | 17 - 18 | 2898 | 112 | ||
BC061202.1 | - | 17 - 18 | 2878 | 112 | ||
BC138734.1 | - | 17 - 18 | 2944 | 8861756 | 112 | |
BC138735.1 | - | 17 - 18 | 2944 | 8861757 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |