Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028639.3 | NP_082915.1 | 13 - 14 | 1911 | 82 | |
GenBank mRNA | AK040578.1 | - | 13 - 14 | 1914 | 82 | |
AK040610.1 | - | 13 - 14 | 1882 | 82 | ||
AK050263.1 | - | 13 - 14 | 1898 | 82 | ||
AK080284.1 | - | 13 - 14 | 1890 | 82 | ||
AK153704.1 | - | 13 - 14 | 1893 | 82 | ||
AK165244.1 | - | 6 - 7 | 573 | 82 | ||
BC023773.1 | - | 9 - 10 | 869 | 82 | ||
BC042512.1 | - | 13 - 14 | 1588 | 4223801 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |