Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081424.1 | NP_001074893.1 | 3 - 4 | 755 | 85 | |
GenBank mRNA | AK033068.1 | - | 3 - 4 | 755 | 85 | |
BC147245.1 | - | 3 - 4 | 470 | 9056293 | 85 | |
BC147246.1 | - | 3 - 4 | 470 | 9056310 | 85 | |
BC151033.1 | - | 3 - 4 | 470 | 9087946 | 85 | |
BC151035.1 | - | 3 - 4 | 470 | 9087948 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |