Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144825.2 | NP_659074.2 | 9 - 10 | 1449 | 87 | |
GenBank mRNA | AK138443.1 | - | 1421 | 87 | ||
AK143530.1 | - | 9 - 10 | 1456 | 87 | ||
BC047271.1 | - | 8 - 9 | 796 | 87 | ||
BC151012.1 | - | 9 - 10 | 1243 | 9087925 | 87 | |
BC151015.1 | - | 9 - 10 | 1243 | 9087928 | 87 | |
BC151016.1 | - | 9 - 10 | 1242 | 9087929 | 87 | |
CN724017.1 | - | 1 - 2 | 43 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |