Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571065 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571068 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571067 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571064 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2073294 |
AK168594.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2073294 |
AK169637.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2073294 |
BC005585.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2073294 |
AK083164.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2073294 |
AK083767.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809127 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671902 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770893 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
421711 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
963084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501758 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777375 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539571 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274227 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380968 |
AF033017.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380968 |
AK005326.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380968 |
AK137224.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380968 |
BC003729.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
380968 |
NM_008430.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633460 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
618994 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
511076 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
23748 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639064 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562605 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81125 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
357392 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
25395 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176722 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
149470 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123147 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P132097 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073294 |
AK168594.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073294 |
AK169637.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073294 |
BC005585.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073294 |
AK083164.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073294 |
AK083767.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P217990 |
AK033462.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571092 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571077 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571083 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571065 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571072 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571075 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571088 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571069 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571068 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571078 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571073 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571086 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571064 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23571081 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05519548 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01064158 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P132097 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123147 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05519533 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P217990 |
AK033462.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118290 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109912 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130187 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123147 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P132097 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04983735 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04983741 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571065 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571067 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571068 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23571064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23571263 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571090 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571118 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571109 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04983745 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04781376 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04983744 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571110 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571088 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04983749 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571069 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571221 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04532538 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571113 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571216 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571116 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571215 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571219 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571073 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571114 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571077 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571102 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571111 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04532542 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571218 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571099 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571220 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23571119 |
- |
- |