Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028149.1 | NP_082425.1 | 8 - 9 | 885 | 107 | |
GenBank mRNA | AK012109.1 | - | 8 - 9 | 609 | 107 | |
AK144786.1 | - | 8 - 9 | 859 | 107 | ||
AK220232.1 | - | 8 - 9 | 838 | 107 | ||
EF649694.1 | - | 8 - 9 | 632 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |