Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100023020.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023020.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023020.mm.1 |
TC1100002471.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023017.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023017.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023017.mm.1 |
TC1100002471.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023016.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023016.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023016.mm.1 |
TC1100002471.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023019.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023019.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023019.mm.1 |
TC1100002471.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023021.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023021.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023021.mm.1 |
TC1100002471.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100023024.mm.1 |
NM_010117.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023024.mm.1 |
NM_010117.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100023024.mm.1 |
TC1100002471.mm.1 |
MTA TC |