Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011574.2 | NP_035704.2 | 13 - 14 | 1632 | 75 | |
GenBank mRNA | AK144455.1 | - | 13 - 14 | 1617 | 75 | |
AK146263.1 | - | 13 - 14 | 1608 | 75 | ||
AK150862.1 | - | 13 - 14 | 1607 | 75 | ||
AK168470.1 | - | 13 - 14 | 1582 | 75 | ||
AK173318.1 | - | 12 - 13 | 1573 | 75 | ||
BC018262.1 | - | 1 - 2 | 85 | 75 | ||
BC027399.1 | - | 9 - 10 | 1131 | 4948207 | 75 | |
BC068113.1 | - | 13 - 14 | 1621 | 5707865 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |