Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028385.1 | NP_082661.1 | 14 - 15 | 2284 | 80 | |
GenBank mRNA | AK049143.1 | - | 8 - 9 | 1362 | 80 | |
AK122551.1 | - | 14 - 15 | 2284 | 80 | ||
BC056502.1 | - | 17 - 18 | 2623 | 80 | ||
BC083184.1 | - | 13 - 14 | 2165 | 30536831 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |