Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028882.4 | NP_083158.3 | 10 - 11 | 1597 | 126 | |
GenBank mRNA | AK028900.1 | - | 10 - 11 | 1379 | 126 | |
AK031704.1 | - | 10 - 11 | 1209 | 126 | ||
AK051165.1 | - | 10 - 11 | 126 | |||
AK052671.1 | - | 10 - 11 | 1598 | 126 | ||
AK153839.1 | - | 10 - 11 | 1258 | 126 | ||
BC138131.1 | - | 10 - 11 | 1140 | 8861150 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |