Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173865.3 | NP_776290.1 | 3 - 4 | 1469 | 83 | |
GenBank mRNA | AK089802.1 | - | 3 - 4 | 1470 | 83 | |
BC096596.1 | - | 3 - 4 | 1505 | 4239523 | 83 | |
BC116280.1 | - | 3 - 4 | 1277 | 40094408 | 83 | |
BC116281.1 | - | 3 - 4 | 1277 | 40094411 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |