Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173865.3 | NP_776290.1 | 4 - 5 | 1543 | 73 | |
GenBank mRNA | AK089802.1 | - | 4 - 5 | 1544 | 73 | |
AK154819.1 | - | 4 - 5 | 1517 | 73 | ||
BC096596.1 | - | 4 - 5 | 1579 | 4239523 | 73 | |
BC116280.1 | - | 4 - 5 | 1351 | 40094408 | 73 | |
BC116281.1 | - | 4 - 5 | 1351 | 40094411 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |