Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174847.2 | NP_777272.2 | 6 - 7 | 1149 | 98 | |
GenBank mRNA | AK017896.1 | - | 6 - 7 | 1134 | 98 | |
AK077731.1 | - | 6 - 7 | 1110 | 98 | ||
AK080956.1 | - | 6 - 7 | 1093 | 98 | ||
AK086520.1 | - | 6 - 7 | 1151 | 98 | ||
AK143875.1 | - | 6 - 7 | 1123 | 98 | ||
AK169902.1 | - | 6 - 7 | 1085 | 98 | ||
AK220454.1 | - | 6 - 7 | 1134 | 98 | ||
BC043688.1 | - | 6 - 7 | 1110 | 5136661 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |