Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mael | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175296.4 | NP_780505.1 | 3 - 4 | 402 | 141 | |
GenBank mRNA | AK077103.1 | - | 3 - 4 | 399 | 141 | |
AK165801.1 | - | 3 - 4 | 399 | 141 | ||
BC050800.1 | - | 3 - 4 | 402 | 6744032 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |