Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242407.1 | NP_001229336.1 | 12 - 13 | 1484 | 75 | |
NM_175392.3 | NP_780601.1 | 12 - 13 | 1652 | 75 | ||
GenBank mRNA | AK077618.1 | - | 12 - 13 | 1445 | 75 | |
AK131152.1 | - | 9 - 10 | 2334 | 75 | ||
AK154200.1 | - | 12 - 13 | 1473 | 75 | ||
AK165996.1 | - | 12 - 13 | 1476 | 75 | ||
BC051404.1 | - | 12 - 13 | 1652 | 6395030 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |