Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301204.1 | NP_001288133.1 | 6 - 7 | 908 | 93 | |
NM_001301205.1 | NP_001288134.1 | 5 - 6 | 593 | 93 | ||
NM_176913.4 | NP_795887.2 | 6 - 7 | 1032 | 93 | ||
GenBank mRNA | AF488552.1 | - | 6 - 7 | 1032 | 93 | |
AK089165.1 | - | 3 - 4 | 2646 | 93 | ||
AK089244.1 | - | 6 - 7 | 894 | 93 | ||
AK155099.1 | - | 6 - 7 | 860 | 93 | ||
AK155308.1 | - | 6 - 7 | 859 | 93 | ||
AK155417.1 | - | 6 - 7 | 863 | 93 | ||
AK170094.1 | - | 6 - 7 | 860 | 93 | ||
AK170970.1 | - | 6 - 7 | 860 | 93 | ||
AK171044.1 | - | 6 - 7 | 860 | 93 | ||
AK171122.1 | - | 6 - 7 | 860 | 93 | ||
AK171155.1 | - | 5 - 6 | 573 | 93 | ||
AK171196.1 | - | 6 - 7 | 860 | 93 | ||
AK171268.1 | - | 6 - 7 | 860 | 93 | ||
AK171503.1 | - | 6 - 7 | 860 | 93 | ||
AK171606.1 | - | 6 - 7 | 876 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Detect Genomic DNA Mus musculus Probe spans exons |