Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177068.4 | NP_796042.3 | 4 - 5 | 1118 | 105 | |
GenBank mRNA | AK029292.1 | - | 4 - 5 | 1090 | 105 | |
AK042424.1 | - | 4 - 5 | 1119 | 105 | ||
AK132477.1 | - | 4 - 5 | 828 | 105 | ||
AK172136.1 | - | 4 - 5 | 1083 | 105 | ||
AK172499.1 | - | 4 - 5 | 1083 | 105 | ||
BC025654.1 | - | 4 - 5 | 828 | 5342879 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |