Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28237416 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28237415 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
245784 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588330 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
337863 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
657180 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306975 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
200650 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
846664 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
178421 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
660348 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
482914 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970639 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
193580 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
573097 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
778699 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
25910 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
AK006827.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
AK045082.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
AK137080.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
AK137471.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
AK164465.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
BC026785.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
BC057940.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
L29479.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
L29480.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305089 |
NM_011495.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
51989 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
117485 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11861 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P171453 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P107413 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237388 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237383 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237403 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237385 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237413 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237395 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237397 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237398 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237391 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237389 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237408 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237412 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237399 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237409 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237380 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237382 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237407 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237405 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237415 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237400 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237410 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237394 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28237387 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07538673 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P107413 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02461004 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P171453 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P661451 |
BC138157.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P661451 |
BC138158.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P661451 |
AK158255.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P661451 |
BC090653.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P661451 |
BC099936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
BC090653.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
BC099936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
AK030995.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
AK031077.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P64763 |
NM_177331.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P638 |
AK045081.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P205951 |
BC099936.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P205951 |
AK031077.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P120323 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07538673 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P107413 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P171453 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09902353 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09902355 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09902359 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28237415 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28237416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237291 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237319 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237298 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28237405 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237347 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237397 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237374 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237358 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237371 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10051525 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237305 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10356101 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237228 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237357 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237300 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237352 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237379 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237375 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237383 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237304 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237409 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237313 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237412 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10356096 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10382237 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237238 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237246 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237373 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237239 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237223 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237249 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237226 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237345 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237408 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237235 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237392 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237231 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10028549 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237343 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237224 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237229 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237391 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10028550 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237253 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237241 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237225 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237378 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237227 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09902355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237233 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237405 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237400 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237311 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237356 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10395649 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237301 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237380 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P10422209 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237245 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237252 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28237401 |
- |
- |