Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178633.3 | NP_848748.2 | 1501 | 105 | ||
GenBank mRNA | AK020050.1 | - | 152 | 105 | ||
AK032116.1 | - | 1049 | 105 | |||
AK167903.1 | - | 1292 | 105 | |||
AK208170.1 | - | 189 | 105 | |||
AK220570.1 | - | 1501 | 105 | |||
BC024572.1 | - | 668 | 105 | |||
BC031142.1 | - | 1372 | 4972965 | 105 | ||
BC031144.1 | - | 1395 | 4973325 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |