Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178635.3 | NP_848750.3 | 3 - 4 | 468 | 132 | |
GenBank mRNA | AK030814.1 | - | 3 - 4 | 399 | 132 | |
AK043084.1 | - | 3 - 4 | 446 | 132 | ||
AK044129.1 | - | 3 - 4 | 451 | 132 | ||
AK078085.1 | - | 3 - 4 | 460 | 132 | ||
AK089900.1 | - | 3 - 4 | 453 | 132 | ||
AK154903.1 | - | 3 - 4 | 468 | 132 | ||
AK165015.1 | - | 3 - 4 | 456 | 132 | ||
AK165185.1 | - | 3 - 4 | 448 | 132 | ||
BC034176.1 | - | 3 - 4 | 437 | 4954871 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |