Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181517.3 | NP_852658.2 | 13 - 14 | 1430 | 142 | |
GenBank mRNA | AK039534.1 | - | 13 - 14 | 1582 | 142 | |
AK079903.1 | - | 12 - 13 | 2940 | 142 | ||
AY316153.1 | - | 13 - 14 | 1430 | 142 | ||
BC053524.1 | - | 10 - 11 | 1113 | 142 | ||
BC064825.1 | - | 4 - 5 | 386 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |