Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624112 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624113 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624114 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624111 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496760 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
16956 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
791473 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
30335 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
699646 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
232280 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
53260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
74553 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AF175771.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AF291425.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AJ279791.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AJ279792.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AJ279793.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AJ279794.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AJ279795.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AK013248.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AK017989.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AK041748.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AK154694.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AK171199.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
AY039000.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
BC051204.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
BC055858.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
882394 |
NM_020286.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
887593 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
727723 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304580 |
AK081822.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
102022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
141415 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302428 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
271673 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
142510 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
134237 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102621 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127207 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624124 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624109 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624107 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624105 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21624120 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P127207 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04689648 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102621 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04689597 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04689625 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P399584 |
CV560846.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P399584 |
BC053443.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P399584 |
NM_181589.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140389 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134831 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119982 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P127207 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102621 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01343591 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01343589 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01343594 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01343590 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01521093 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624113 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624111 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624112 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21624114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623995 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21623896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21624172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623990 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623964 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01623096 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624093 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623915 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623900 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623962 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624122 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624089 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624101 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623983 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624110 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01235285 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623970 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624091 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624124 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623916 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623961 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623890 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624100 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01925330 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624118 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01343593 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623987 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02215741 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624105 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623913 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623966 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624112 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623954 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01623087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623903 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01343594 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623892 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624106 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624120 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624107 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623889 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01623084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01213443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623950 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624096 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01623083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21623989 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624108 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624094 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01607947 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624114 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21624103 |
- |
- |