Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181728.2 | NP_859417.2 | 3 - 4 | 282 | 64 | |
GenBank mRNA | AJ311769.1 | - | 1 - 2 | 66 | 64 | |
AJ311770.1 | - | 1 - 2 | 66 | 64 | ||
AJ311771.1 | - | 1 - 2 | 66 | 64 | ||
AJ311772.1 | - | 1 - 2 | 66 | 64 | ||
AJ311773.1 | - | 1 - 2 | 66 | 64 | ||
AK016257.1 | - | 3 - 4 | 283 | 64 | ||
BC141069.1 | - | 1 - 2 | 142 | 9055681 | 64 | |
BC145327.1 | - | 1 - 2 | 142 | 9053883 | 64 | |
DQ519368.1 | - | 3 - 4 | 281 | 64 | ||
DQ519369.1 | - | 3 - 4 | 230 | 64 | ||
DQ519370.1 | - | 2 - 3 | 159 | 64 | ||
DQ519371.1 | - | 1 - 2 | 358 | 64 | ||
DQ810295.1 | - | 2 - 3 | 146 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |