Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181730.4 | NP_859419.1 | 1 - 2 | 349 | 68 | |
GenBank mRNA | AK003353.1 | - | 1 - 2 | 128 | 68 | |
AK154241.1 | - | 1 - 2 | 124 | 68 | ||
BC002025.1 | - | 1 - 2 | 114 | 3591169 | 68 | |
BC024394.1 | - | 1 - 2 | 114 | 4987975 | 68 | |
BC087884.1 | - | 1 - 2 | 148 | 6772717 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |