Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170912.1 | NP_001164383.1 | 18 - 19 | 3939 | 60 | |
NM_001170913.1 | NP_001164384.1 | 18 - 19 | 3977 | 60 | ||
NM_181853.4 | NP_862901.3 | 18 - 19 | 3876 | 60 | ||
GenBank mRNA | AK147559.1 | - | 18 - 19 | 3978 | 60 | |
AK172929.1 | - | 3 - 4 | 350 | 60 | ||
AY572454.1 | - | 18 - 19 | 3939 | 60 | ||
AY572455.1 | - | 18 - 19 | 3876 | 60 | ||
BC138032.1 | - | 17 - 18 | 3549 | 8861051 | 60 | |
BC145106.1 | - | 17 - 18 | 3549 | 9053654 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |