Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117832 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117834 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117831 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117829 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117833 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117830 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817622 |
AK028047.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817622 |
BC006844.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817622 |
BC031768.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817622 |
NM_177614.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
184302 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
959602 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
377543 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
717906 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
864719 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
605941 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
362800 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
48482 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168385 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P151425 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117854 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117831 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117834 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117840 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117833 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117860 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117842 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117848 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117838 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117855 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117829 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24117857 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168385 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01229107 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P151425 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
M16229.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK135162.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK160553.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
AK167809.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P384879 |
BC023482.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136845 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P151425 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168385 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04807435 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04807449 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04807434 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117830 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117829 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117831 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117833 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117832 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24117834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24117874 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04271276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04729239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117860 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117846 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04807443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04496684 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117866 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04321740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117848 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04807435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04321739 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117831 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117855 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117857 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117849 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04884785 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117853 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117829 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117852 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117865 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04703268 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04194331 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117845 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24117838 |
- |
- |