Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003548.1 | 2 | 568 | 113 | ||
GenBank mRNA | AY158991.1 | - | 2 | 567 | 113 | |
BC130233.1 | - | 2 | 551 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon |