Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027475.3 | NP_081751.1 | 1 - 2 | 326 | 68 | |
GenBank mRNA | AK028664.1 | - | 1 - 2 | 290 | 68 | |
AK029169.1 | - | 1 - 2 | 258 | 68 | ||
AK043261.1 | - | 1 - 2 | 175 | 68 | ||
BC013470.1 | - | 1 - 2 | 275 | 4220504 | 68 | |
BY272088.1 | - | 1 - 2 | 327 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |