Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146244.4 | NP_666356.2 | 7 - 8 | 1380 | 68 | |
GenBank mRNA | AK044050.1 | - | 7 - 8 | 1371 | 68 | |
BC027298.1 | - | 7 - 8 | 1334 | 3672127 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |