Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011069.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011069.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011069.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011069.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011069.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011070.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011070.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011070.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011070.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011070.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011072.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011072.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011072.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011076.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011076.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011076.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011076.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011076.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011078.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011078.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011078.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011078.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011078.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011067.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011067.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011067.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011067.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011067.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011071.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011071.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011071.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011077.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011077.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011077.mm.1 |
NM_028800.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011077.mm.1 |
NM_028800.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011077.mm.1 |
TC0400001453.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011074.mm.1 |
NM_001145827.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011074.mm.1 |
NM_001145827.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011074.mm.1 |
TC0400001453.mm.1 |
MTA TC |