Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033573.1 | NP_001028745.1 | 4 - 5 | 1074 | 78 | |
NM_025883.3 | NP_080159.3 | 4 - 5 | 958 | 78 | ||
GenBank mRNA | AK010296.1 | - | 4 - 5 | 953 | 78 | |
AK011373.1 | - | 4 - 5 | 925 | 78 | ||
AK035172.1 | - | 1037 | 78 | |||
AK038177.1 | - | 937 | 78 | |||
AK050388.1 | - | 4 - 5 | 1073 | 78 | ||
AK077693.1 | - | 4 - 5 | 912 | 78 | ||
AK077838.1 | - | 923 | 78 | |||
AK139707.1 | - | 4 - 5 | 915 | 78 | ||
AK144128.1 | - | 4 - 5 | 930 | 78 | ||
AK154741.1 | - | 4 - 5 | 936 | 78 | ||
AK157494.1 | - | 4 - 5 | 940 | 78 | ||
AK159304.1 | - | 4 - 5 | 959 | 78 | ||
AK168497.1 | - | 5 - 6 | 1081 | 78 | ||
AK183417.1 | - | 2 - 3 | 187 | 78 | ||
AK184113.1 | - | 2 - 3 | 186 | 78 | ||
AK210114.1 | - | 2 - 3 | 186 | 78 | ||
AK214916.1 | - | 2 - 3 | 186 | 78 | ||
BC033317.1 | - | 4 - 5 | 905 | 4459242 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |