Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020011.5 | NP_064395.2 | 1 - 2 | 611 | 85 | |
GenBank mRNA | AK004951.1 | - | 1 - 2 | 611 | 85 | |
BC092084.1 | - | 1 - 2 | 60 | 4987298 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |