Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975718 |
AV252647.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975718 |
EU099306.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975718 |
NM_001142775.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1975718 |
AK132639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2120388 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
297710 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
598173 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
850815 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385518 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198301 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
669721 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
869071 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
104087 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356482 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
418706 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658053 |
AK018189.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658053 |
AK220482.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658053 |
BC119557.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658053 |
BC119558.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658053 |
XM_355952.5 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356186 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
194659 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
97887 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
149470 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410473 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
442758 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174292 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
125083 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
104226 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P175796 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P172333 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975718 |
AV252647.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975718 |
EU099306.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975718 |
NM_001142775.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1975718 |
AK132639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2120388 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213392 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213385 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213406 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213382 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213401 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213389 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213386 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213380 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213404 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213400 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23213383 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P175796 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P172333 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P248181 |
NM_177818.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P248181 |
AK048392.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125418 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103295 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P175796 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P172333 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03359631 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03359625 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03359637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23213400 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213415 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213406 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03742609 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213389 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213388 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213374 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213382 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213417 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213400 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213420 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213391 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213373 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213383 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213385 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213421 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213404 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213377 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213401 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213387 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03713592 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03513989 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213408 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03905493 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03713587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03446003 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213375 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213392 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213413 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23213399 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04097293 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03215800 |
- |
- |