Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF196314.1 | - | 546 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus |