Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007592.3 | NP_031618.2 | 2 - 3 | 382 | 138 | |
GenBank mRNA | AK004896.1 | - | 2 - 3 | 383 | 138 | |
AK004923.1 | - | 2 - 3 | 383 | 138 | ||
BC010773.1 | - | 2 - 3 | 358 | 4216498 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |