Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 39 - 40 | 2896 | 78 | |
GenBank mRNA | AK132180.1 | - | 39 - 40 | 2917 | 78 | |
AK159283.1 | - | 39 - 40 | 2914 | 78 | ||
AK159285.1 | - | 39 - 40 | 2918 | 78 | ||
BC003198.1 | - | 2 - 3 | 206 | 78 | ||
BC042642.1 | - | 32 - 33 | 2466 | 78 | ||
BC050014.1 | - | 39 - 40 | 2896 | 4225038 | 78 | |
M14423.1 | - | 17 - 18 | 1246 | 78 | ||
U08020.1 | - | 39 - 40 | 2797 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |