Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 40 - 41 | 3005 | 61 | |
GenBank mRNA | AK132180.1 | - | 40 - 41 | 3026 | 61 | |
AK159283.1 | - | 40 - 41 | 3023 | 61 | ||
AK159285.1 | - | 40 - 41 | 3027 | 61 | ||
BC003198.1 | - | 3 - 4 | 315 | 61 | ||
BC042642.1 | - | 33 - 34 | 2575 | 61 | ||
BC050014.1 | - | 40 - 41 | 3005 | 4225038 | 61 | |
M14423.1 | - | 18 - 19 | 1355 | 61 | ||
U08020.1 | - | 40 - 41 | 2906 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |