Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281819.1 | NP_001268748.1 | 23 - 24 | 3553 | 67 | |
NM_009598.2 | NP_033728.1 | 12 - 13 | 1776 | 67 | ||
NM_207624.5 | NP_997507.1 | 23 - 24 | 3553 | 67 | ||
GenBank mRNA | AK154632.1 | - | 23 - 24 | 3552 | 67 | |
AK161020.1 | - | 23 - 24 | 3549 | 67 | ||
AK164780.1 | - | 10 - 11 | 1421 | 67 | ||
BC034367.1 | - | 19 - 20 | 2815 | 67 | ||
BC040404.1 | - | 23 - 24 | 3542 | 3965649 | 67 | |
BC052531.1 | - | 4 - 5 | 405 | 67 | ||
BC083109.1 | - | 23 - 24 | 3331 | 67 | ||
BC110362.1 | - | 23 - 24 | 3539 | 5343978 | 67 | |
J04946.1 | - | 17 - 18 | 2520 | 67 | ||
J04947.1 | - | 2520 | 67 | |||
M55333.1 | - | 12 - 13 | 1776 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |