Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004002.1 | - | 2 - 3 | 243 | 74 | |
AK075967.1 | - | 2 - 3 | 226 | 74 | ||
BC026209.1 | - | 2 - 3 | 230 | 1349819 | 74 | |
M96554.1 | - | 2 - 3 | 175 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |