Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600014073.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014073.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014073.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014071.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014071.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014071.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014078.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014078.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014078.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014075.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014075.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014075.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014079.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014079.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014079.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014072.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014072.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014072.mm.1 |
NM_009753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014074.mm.1 |
TC0600001775.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600014074.mm.1 |
NM_001112796.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600014074.mm.1 |
NM_009753.4 |
Refseq |