Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009968.3 | NP_034098.1 | 1 - 2 | 567 | 89 | |
GenBank mRNA | AK098082.1 | - | 1 - 2 | 51 | 89 | |
AK133820.1 | - | 1 - 2 | 568 | 89 | ||
AK134281.1 | - | 1 - 2 | 52 | 89 | ||
BC003800.1 | - | 1 - 2 | 59 | 3492859 | 89 | |
S70056.1 | - | 1 - 2 | 79 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |