Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010258.3 | NP_034388.2 | 2 - 3 | 1387 | 120 | |
GenBank mRNA | AF179425.1 | - | 1 - 2 | 694 | 120 | |
AK142381.1 | - | 2 - 3 | 1247 | 120 | ||
S82462.1 | - | 1 - 2 | 1149 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |