Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010273.4 | NP_034403.1 | 9 - 10 | 1519 | 91 | |
GenBank mRNA | AF251042.1 | - | 9 - 10 | 1188 | 91 | |
AK028880.1 | - | 9 - 10 | 1276 | 91 | ||
AK032869.1 | - | 8 - 9 | 1525 | 91 | ||
AK156224.1 | - | 9 - 10 | 1287 | 91 | ||
AK171216.1 | - | 9 - 10 | 1276 | 91 | ||
BC010220.1 | - | 1 - 2 | 73 | 91 | ||
BC013758.1 | - | 9 - 10 | 1200 | 4501084 | 91 | |
BC025606.1 | - | 9 - 10 | 1286 | 91 | ||
BC037598.1 | - | 9 - 10 | 1253 | 5249271 | 91 | |
U07950.1 | - | 5 - 6 | 762 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |