Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010616.2 | NP_034746.1 | 5 - 6 | 492 | 95 | |
GenBank mRNA | AB001428.1 | - | 1 - 2 | 69 | 95 | |
AB755406.1 | - | 5 - 6 | 505 | 95 | ||
AK018598.1 | - | 5 - 6 | 494 | 95 | ||
BC022225.1 | - | 5 - 6 | 422 | 4222522 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |