Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011030.2 | NP_035160.1 | 3 - 4 | 386 | 78 | |
GenBank mRNA | AK030574.1 | - | 3 - 4 | 318 | 78 | |
AK045008.1 | - | 3 - 4 | 315 | 78 | ||
AK149127.1 | - | 3 - 4 | 278 | 78 | ||
AK160798.1 | - | 3 - 4 | 315 | 78 | ||
AK161212.1 | - | 3 - 4 | 319 | 78 | ||
AK165468.1 | - | 3 - 4 | 315 | 78 | ||
AK169726.1 | - | 3 - 4 | 319 | 78 | ||
BC009654.1 | - | 3 - 4 | 309 | 3484815 | 78 | |
BE985798.1 | - | 3 - 4 | 386 | 78 | ||
BY721363.1 | - | 3 - 4 | 367 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |