Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900012777.mm.1 |
NM_001159487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012777.mm.1 |
NM_001159487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012777.mm.1 |
NM_011255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012777.mm.1 |
NM_011255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012777.mm.1 |
TC1900001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012781.mm.1 |
NM_001159487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012781.mm.1 |
NM_001159487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012781.mm.1 |
TC1900001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012782.mm.1 |
NM_001159487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012782.mm.1 |
NM_001159487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012782.mm.1 |
TC1900001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012778.mm.1 |
NM_001159487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012778.mm.1 |
NM_001159487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012778.mm.1 |
NM_011255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012778.mm.1 |
NM_011255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012778.mm.1 |
TC1900001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012776.mm.1 |
NM_001159487.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012776.mm.1 |
NM_001159487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012776.mm.1 |
NM_011255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012776.mm.1 |
NM_011255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012776.mm.1 |
TC1900001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900012779.mm.1 |
NM_011255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012779.mm.1 |
NM_011255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900012779.mm.1 |
TC1900001464.mm.1 |
MTA TC |