Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011578.3 | NP_035708.2 | 6 - 7 | 1169 | 100 | |
GenBank mRNA | AF039601.1 | - | 6 - 7 | 969 | 100 | |
AK166082.1 | - | 6 - 7 | 1170 | 100 | ||
BC070428.1 | - | 6 - 7 | 1094 | 6410554 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |