Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011943.2 | NP_036073.1 | 3 - 4 | 416 | 70 | |
GenBank mRNA | AK043482.1 | - | 3 - 4 | 417 | 70 | |
AK077712.1 | - | 3 - 4 | 411 | 70 | ||
AK086722.1 | - | 417 | 70 | |||
AK138757.1 | - | 3 - 4 | 417 | 70 | ||
AK140289.1 | - | 4 - 5 | 401 | 70 | ||
AK141708.1 | - | 3 - 4 | 401 | 70 | ||
AK162671.1 | - | 3 - 4 | 370 | 70 | ||
BC075652.1 | - | 3 - 4 | 415 | 30541969 | 70 | |
U39066.1 | - | 3 - 4 | 330 | 70 | ||
X97052.1 | - | 3 - 4 | 274 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |