Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012002.3 | NP_036132.1 | 8 - 9 | 745 | 79 | |
GenBank mRNA | AF071315.1 | - | 8 - 9 | 782 | 79 | |
AK146410.1 | - | 8 - 9 | 756 | 79 | ||
AK146775.1 | - | 8 - 9 | 758 | 79 | ||
BC004664.1 | - | 8 - 9 | 744 | 3498214 | 79 | |
BC014286.1 | - | 8 - 9 | 749 | 4163277 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |