Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012055.3 | NP_036185.1 | 2 - 3 | 344 | 126 | |
GenBank mRNA | AK076207.1 | - | 2 - 3 | 362 | 126 | |
AK160459.1 | - | 2 - 3 | 359 | 126 | ||
AK167524.1 | - | 2 - 3 | 324 | 126 | ||
AK167933.1 | - | 2 - 3 | 343 | 126 | ||
BC005552.1 | - | 2 - 3 | 342 | 3585775 | 126 | |
BC050123.1 | - | 2672 | 126 | |||
U38940.1 | - | 2 - 3 | 345 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |